siRNA Details
Gene Symbol: Ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127531.2 | NP_001121003.1 | 5 | 570 | ||
XM_006256860.2 | XP_006256922.1 | 6 | 651 | |||
GenBank mRNA | BC166480.1 | - | 5 | 570 | 9102341 | |
FQ235170.1 | - | 2 | 65 |
