siRNA Details
| Gene Symbol: Rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104643.1 | NP_001098113.1 | 4, 5 | 334 | ||
| GenBank mRNA | EU074986.1 | - | 4, 5 | 334 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rs1| Gene Symbol: Rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104643.1 | NP_001098113.1 | 4, 5 | 334 | ||
| GenBank mRNA | EU074986.1 | - | 4, 5 | 334 | ||