siRNA Details
| Gene Symbol: Klhl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_228962.8 | XP_228962.2 | 1 | 1703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl34| Gene Symbol: Klhl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_228962.8 | XP_228962.2 | 1 | 1703 | ||