siRNA Details
Gene Symbol: Igsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175763.2 | NP_786939.1 | 6 | 832 | ||
XM_006257557.2 | XP_006257619.1 | 7 | 926 | |||
XM_006257558.1 | XP_006257620.1 | 6 | 876 | |||
XM_006257559.1 | XP_006257621.1 | 6 | 879 | |||
XM_006257560.1 | XP_006257622.1 | 6 | 879 | |||
XM_006257561.1 | XP_006257623.1 | 6 | 879 | |||
XM_008773563.1 | XP_008771785.1 | 6 | 879 | |||
GenBank mRNA | AF322216.1 | - | 6 | 832 |
