siRNA Details
| Gene Symbol: Gpr112l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773667.1 | XP_008771889.1 | 3 | 3892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr112l| Gene Symbol: Gpr112l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773667.1 | XP_008771889.1 | 3 | 3892 | ||