siRNA Details
| Gene Symbol: Slc9a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773665.1 | XP_008771887.1 | 5 | 616 | ||
| XM_217630.8 | XP_217630.3 | 4 | 656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a6| Gene Symbol: Slc9a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773665.1 | XP_008771887.1 | 5 | 616 | ||
| XM_217630.8 | XP_217630.3 | 4 | 656 | |||