siRNA Details
| Gene Symbol: Anks3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009676.1 | NP_001009676.1 | 4 | 466 | ||
| XM_006245788.1 | XP_006245850.1 | 4 | 509 | |||
| XM_006245789.2 | XP_006245851.1 | 4 | 509 | |||
| XM_006245790.1 | XP_006245852.1 | 4 | 402 | |||
| XM_008767482.1 | XP_008765704.1 | 4 | 509 | |||
| XM_008767483.1 | XP_008765705.1 | 3 | 2238 | |||
| XM_008767484.1 | XP_008765706.1 | 2 | 113 | |||
| GenBank mRNA | BC087062.1 | - | 4 | 466 | 7131099 | |

Anks3
Genomic Map