siRNA Details
| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277154.1 | NP_001264083.1 | 12 | 7444 | ||
| GenBank mRNA | BC166455.1 | - | 1 | 636 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srrm2| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277154.1 | NP_001264083.1 | 12 | 7444 | ||
| GenBank mRNA | BC166455.1 | - | 1 | 636 | ||