siRNA Details
| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106984.1 | NP_001100454.1 | 3 | 265 | ||
| GenBank mRNA | AJ005642.1 | - | 2 | 249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss22| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106984.1 | NP_001100454.1 | 3 | 265 | ||
| GenBank mRNA | AJ005642.1 | - | 2 | 249 | ||