siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246057.2 | XP_006246119.1 | 13 | 1801 | ||
| XM_006246059.2 | XP_006246121.1 | 3 | 2603 | |||
| XM_008767609.1 | XP_008765831.1 | 12 | 1573 | |||
| XM_008767610.1 | XP_008765832.1 | 13 | 1711 | |||
| XM_008767611.1 | XP_008765833.1 | 13 | 1652 | |||
| XM_008767612.1 | XP_008765834.1 | 9 | 1541 | |||
| XM_008767613.1 | XP_008765835.1 | 4 | 2831 | |||
| GenBank mRNA | FQ211597.1 | - | 12 | 1540 | ||

Unkl
Genomic Map