siRNA Details
Gene Symbol: Unkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246057.2 | XP_006246119.1 | 12, 13 | 1606 | ||
XM_008767610.1 | XP_008765832.1 | 12, 13 | 1516 | |||
XM_008767611.1 | XP_008765833.1 | 12, 13 | 1457 | |||
XM_008767612.1 | XP_008765834.1 | 8, 9 | 1346 | |||
XM_008767613.1 | XP_008765835.1 | 3, 4 | 2636 |
