siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246057.2 | XP_006246119.1 | 15 | 2162 | ||
| XM_006246059.2 | XP_006246121.1 | 5 | 2964 | |||
| XM_008767609.1 | XP_008765831.1 | 14 | 1934 | |||
| XM_008767610.1 | XP_008765832.1 | 15 | 2072 | |||
| XM_008767611.1 | XP_008765833.1 | 15 | 2013 | |||
| XM_008767612.1 | XP_008765834.1 | 11 | 1902 | |||
| XM_008767613.1 | XP_008765835.1 | 6 | 3192 | |||
| GenBank mRNA | FQ211597.1 | - | 14 | 1901 | ||

Unkl
Genomic Map