siRNA Details
| Gene Symbol: LOC302999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750793.3 | XP_003750841.1 | 1 | 632 | ||
| XM_006246070.2 | XP_006246132.1 | 2 | 1320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC302999| Gene Symbol: LOC302999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750793.3 | XP_003750841.1 | 1 | 632 | ||
| XM_006246070.2 | XP_006246132.1 | 2 | 1320 | |||