siRNA Details
Gene Symbol: Crebrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277157.1 | NP_001264086.1 | 6, 7 | 1850 | ||
XM_008767581.1 | XP_008765803.1 | 5, 6 | 2070 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crebrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277157.1 | NP_001264086.1 | 6, 7 | 1850 | ||
XM_008767581.1 | XP_008765803.1 | 5, 6 | 2070 |