siRNA Details
| Gene Symbol: Thg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013966.1 | NP_001013988.1 | 2 | 224 | ||
| GenBank mRNA | BC087596.1 | - | 2 | 224 | 7310343 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thg1l| Gene Symbol: Thg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013966.1 | NP_001013988.1 | 2 | 224 | ||
| GenBank mRNA | BC087596.1 | - | 2 | 224 | 7310343 | |