siRNA Details
| Gene Symbol: Cyfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106996.1 | NP_001100466.1 | 18, 19 | 2165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyfip2| Gene Symbol: Cyfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106996.1 | NP_001100466.1 | 18, 19 | 2165 | ||