siRNA Details
| Gene Symbol: Jade2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106998.1 | NP_001100468.1 | 12 | 2351 | ||
| XM_006246254.2 | XP_006246316.2 | 12 | 3237 | |||
| XM_006246257.2 | XP_006246319.1 | 12 | 2695 | |||
| XM_006246258.2 | XP_006246320.1 | 12 | 2396 | |||
| XM_006246259.2 | XP_006246321.1 | 12 | 3050 | |||
| XM_006246260.2 | XP_006246322.1 | 12 | 2744 | |||
| XM_008767682.1 | XP_008765904.1 | 11 | 5929 | |||

Jade2
Genomic Map