siRNA Details
| Gene Symbol: Aff4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107001.1 | NP_001100471.1 | 13, 14 | 2922 | ||
| XM_006246263.2 | XP_006246325.1 | 13, 14 | 2995 | |||
| XM_006246264.2 | XP_006246326.1 | 12, 13 | 2831 | |||
| XM_008767684.1 | XP_008765906.1 | 13, 14 | 3010 | |||

Aff4
Genomic Map