siRNA Details
| Gene Symbol: LOC303140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767743.1 | XP_008765965.1 | 7 | 1885 | ||
| GenBank mRNA | DQ119106.1 | - | 5 | 1102 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC303140| Gene Symbol: LOC303140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767743.1 | XP_008765965.1 | 7 | 1885 | ||
| GenBank mRNA | DQ119106.1 | - | 5 | 1102 | ||