siRNA Details
| Gene Symbol: LOC303140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767743.1 | XP_008765965.1 | 3 | 1367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC303140| Gene Symbol: LOC303140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767743.1 | XP_008765965.1 | 3 | 1367 | ||