siRNA Details
| Gene Symbol: Slc36a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024764.1 | NP_001019935.1 | 7 | 739 | ||
| XM_008767685.1 | XP_008765907.1 | 8 | 719 | |||
| GenBank mRNA | BC097463.1 | - | 7 | 739 | 7457579 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc36a3| Gene Symbol: Slc36a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024764.1 | NP_001019935.1 | 7 | 739 | ||
| XM_008767685.1 | XP_008765907.1 | 8 | 719 | |||
| GenBank mRNA | BC097463.1 | - | 7 | 739 | 7457579 | |