siRNA Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199390.2 | NP_955422.1 | 6 | 1085 | ||
| XM_006246489.2 | XP_006246551.1 | 6 | 975 | |||
| XM_006246490.2 | XP_006246552.1 | 6 | 1102 | |||
| XM_006246491.1 | XP_006246553.1 | 7 | 1866 | |||
| XM_008767816.1 | XP_008766038.1 | 6 | 978 | |||
| XM_008767817.1 | XP_008766039.1 | 6 | 976 | |||
| GenBank mRNA | AB096213.1 | - | 6 | 968 | ||
| BC085848.1 | - | 6 | 1085 | 7189414 | ||
| FQ212214.1 | - | 6 | 1101 | |||

Flcn
Genomic Map