siRNA Details
| Gene Symbol: Specc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039017.1 | NP_001034106.1 | 4 | 603 | ||
| XM_006246502.2 | XP_006246564.1 | 4 | 662 | |||
| XM_006246503.2 | XP_006246565.1 | 2 | 461 | |||
| XM_008767821.1 | XP_008766043.1 | 4 | 641 | |||
| XM_008767822.1 | XP_008766044.1 | 4 | 946 | |||
| XM_008767823.1 | XP_008766045.1 | 4 | 949 | |||
| GenBank mRNA | BC099105.1 | - | 4 | 603 | 7460369 | |

Specc1
Genomic Map