siRNA Details
| Gene Symbol: Specc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039017.1 | NP_001034106.1 | 4 | 1491 | ||
| XM_006246502.2 | XP_006246564.1 | 4 | 1550 | |||
| XM_006246503.2 | XP_006246565.1 | 2 | 1349 | |||
| XM_008767821.1 | XP_008766043.1 | 4 | 1529 | |||
| XM_008767822.1 | XP_008766044.1 | 4 | 1834 | |||
| XM_008767823.1 | XP_008766045.1 | 4 | 1837 | |||
| GenBank mRNA | BC099105.1 | - | 4 | 1491 | 7460369 | |

Specc1
Genomic Map