siRNA Details
| Gene Symbol: Pipox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012009.1 | NP_001012009.1 | 6 | 879 | ||
| XM_006246958.1 | XP_006247020.1 | 7 | 1015 | |||
| GenBank mRNA | BC088249.1 | - | 6 | 879 | 7370364 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pipox| Gene Symbol: Pipox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012009.1 | NP_001012009.1 | 6 | 879 | ||
| XM_006246958.1 | XP_006247020.1 | 7 | 1015 | |||
| GenBank mRNA | BC088249.1 | - | 6 | 879 | 7370364 | |