siRNA Details
| Gene Symbol: RGD1307929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768148.1 | XP_008766370.1 | 18 | 3519 | ||
| XM_220636.7 | XP_220636.3 | 18 | 3519 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307929| Gene Symbol: RGD1307929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768148.1 | XP_008766370.1 | 18 | 3519 | ||
| XM_220636.7 | XP_220636.3 | 18 | 3519 | |||