siRNA Details
| Gene Symbol: Hic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107021.1 | NP_001100491.1 | 4 | 2218 | ||
| Gene Symbol: LOC100912068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750862.3 | XP_003750910.2 | 2 | 2137 | ||

Hic1
Genomic Map