siRNA Details
| Gene Symbol: Scarf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107022.1 | NP_001100492.1 | 1185 | |||
| GenBank mRNA | BC166488.1 | - | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scarf1| Gene Symbol: Scarf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107022.1 | NP_001100492.1 | 1185 | |||
| GenBank mRNA | BC166488.1 | - | 359 | |||