siRNA Details
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025664.1 | NP_001020835.2 | 4 | 841 | ||
| NM_001042561.1 | NP_001036026.1 | 9 | 1253 | |||
| XM_008768016.1 | XP_008766238.1 | 9 | 2176 | |||
| GenBank mRNA | BC098720.1 | - | 4 | 841 | ||
| BC167746.1 | - | 9 | 1256 | 8364583 | ||

Wsb1
Genomic Map