siRNA Details
Gene Symbol: Rab11fip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107023.1 | NP_001100493.1 | 12 | 1458 | ||
XM_006246903.2 | XP_006246965.1 | 12 | 1441 | |||
XM_006246904.2 | XP_006246966.1 | 10 | 1403 | |||
XM_008768017.1 | XP_008766239.1 | 12 | 1335 |
