siRNA Details
| Gene Symbol: Rhot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107026.2 | NP_001100496.1 | 13, 14 | 1287 | ||
| XM_006246985.2 | XP_006247047.1 | 13, 14 | 1333 | |||
| XM_006246986.1 | XP_006247048.1 | 13, 14 | 1333 | |||
| GenBank mRNA | BC129123.1 | - | 13, 14 | 1287 | 7130774 | |
| FQ211576.1 | - | 13, 14 | 1235 | |||

Rhot1
Genomic Map