siRNA Details
| Gene Symbol: Tbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107033.1 | NP_001100503.1 | 3 | 474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx2| Gene Symbol: Tbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107033.1 | NP_001100503.1 | 3 | 474 | ||