siRNA Details
| Gene Symbol: Med13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107035.1 | NP_001100505.1 | 19 | 4751 | ||
| XM_006247066.2 | XP_006247128.1 | 19 | 4755 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med13| Gene Symbol: Med13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107035.1 | NP_001100505.1 | 19 | 4751 | ||
| XM_006247066.2 | XP_006247128.1 | 19 | 4755 | |||