siRNA Details
| Gene Symbol: Gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044238.1 | NP_001037703.1 | 5 | 606 | ||
| GenBank mRNA | BC105620.1 | - | 5 | 606 | 7453591 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdpd1| Gene Symbol: Gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044238.1 | NP_001037703.1 | 5 | 606 | ||
| GenBank mRNA | BC105620.1 | - | 5 | 606 | 7453591 | |