siRNA Details
Gene Symbol: Gdpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044238.1 | NP_001037703.1 | 7, 8 | 838 | ||
GenBank mRNA | BC105620.1 | - | 7, 8 | 838 | 7453591 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gdpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044238.1 | NP_001037703.1 | 7, 8 | 838 | ||
GenBank mRNA | BC105620.1 | - | 7, 8 | 838 | 7453591 |