siRNA Details
| Gene Symbol: Kat7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181081.2 | NP_851595.1 | 13 | 1795 | ||
| GenBank mRNA | AB334306.1 | - | 12 | 1734 | ||
| AY236854.1 | - | 13 | 1795 | |||
| AY241457.1 | - | 11 | 1660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kat7| Gene Symbol: Kat7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181081.2 | NP_851595.1 | 13 | 1795 | ||
| GenBank mRNA | AB334306.1 | - | 12 | 1734 | ||
| AY236854.1 | - | 13 | 1795 | |||
| AY241457.1 | - | 11 | 1660 | |||