siRNA Details
| Gene Symbol: Ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037643.2 | NP_001032732.2 | 4, 5 | 732 | ||
| GenBank mRNA | BC107663.1 | - | 4, 5 | 732 | 7936709 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2z| Gene Symbol: Ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037643.2 | NP_001032732.2 | 4, 5 | 732 | ||
| GenBank mRNA | BC107663.1 | - | 4, 5 | 732 | 7936709 | |