siRNA Details
Gene Symbol: Arhgap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247437.1 | XP_006247499.1 | 18 | 2970 | ||
XM_006247438.1 | XP_006247500.1 | 18 | 2970 | |||
XM_008768198.1 | XP_008766420.1 | 16 | 3274 | |||
XM_008768199.1 | XP_008766421.1 | 17 | 3181 | |||
XM_008768200.1 | XP_008766422.1 | 16 | 4425 | |||
XM_220918.8 | XP_220918.6 | 20 | 3274 |
