siRNA Details
| Gene Symbol: Plxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107046.1 | NP_001100516.1 | 9 | 1082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxdc1| Gene Symbol: Plxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107046.1 | NP_001100516.1 | 9 | 1082 | ||