siRNA Details
| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107048.1 | NP_001100518.1 | 1, 2 | 806 | ||
| XM_006247304.2 | XP_006247366.1 | 1, 2 | 1125 | |||
| GenBank mRNA | FQ228290.1 | - | 1, 2 | 760 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msl1| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107048.1 | NP_001100518.1 | 1, 2 | 806 | ||
| XM_006247304.2 | XP_006247366.1 | 1, 2 | 1125 | |||
| GenBank mRNA | FQ228290.1 | - | 1, 2 | 760 | ||