siRNA Details
| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107049.1 | NP_001100519.1 | 11 | 1622 | ||
| XM_006247310.2 | XP_006247372.1 | 11 | 2476 | |||
| GenBank mRNA | BC162060.1 | - | 11 | 1565 | 7115660 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc25| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107049.1 | NP_001100519.1 | 11 | 1622 | ||
| XM_006247310.2 | XP_006247372.1 | 11 | 2476 | |||
| GenBank mRNA | BC162060.1 | - | 11 | 1565 | 7115660 | |