siRNA Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213625.1 | NP_998790.1 | 6, 7 | 593 | ||
| XM_006247311.2 | XP_006247373.1 | 7, 8 | 954 | |||
| XM_006247312.2 | XP_006247374.1 | 6, 7 | 674 | |||
| XM_006247313.1 | XP_006247375.1 | 5, 6 | 514 | |||
| GenBank mRNA | AB178470.1 | - | 6, 7 | 593 | ||
| BC093600.1 | - | 5, 6 | 509 | 7316009 | ||
| FQ212948.1 | - | 6, 7 | 624 | |||

Dnajc7
Genomic Map