siRNA Details
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107050.1 | NP_001100520.1 | 11 | 1728 | ||
XM_006247317.2 | XP_006247379.1 | 11 | 1756 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107050.1 | NP_001100520.1 | 11 | 1728 | ||
XM_006247317.2 | XP_006247379.1 | 11 | 1756 |