siRNA Details
| Gene Symbol: Nbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024765.1 | NP_001019936.1 | 11 | 1230 | ||
| XM_006247321.2 | XP_006247383.1 | 11 | 1512 | |||
| XM_006247322.2 | XP_006247384.1 | 11 | 1427 | |||
| XM_006247323.2 | XP_006247385.1 | 11 | 1284 | |||
| XM_006247324.2 | XP_006247386.1 | 11 | 1231 | |||
| XM_006247325.2 | XP_006247387.1 | 11 | 1387 | |||
| XM_006247326.2 | XP_006247388.1 | 11 | 1382 | |||
| GenBank mRNA | BC095903.1 | - | 11 | 1230 | 7096697 | |
| FQ211883.1 | - | 11 | 1230 | |||

Nbr1
Genomic Map