siRNA Details
| Gene Symbol: Lrrc37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768302.1 | XP_008766524.1 | 1 | 2575 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc37a| Gene Symbol: Lrrc37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768302.1 | XP_008766524.1 | 1 | 2575 | ||