siRNA Details
| Gene Symbol: Tmem101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191650.1 | NP_001178579.1 | 2, 3 | 360 | ||
| GenBank mRNA | BC086567.1 | - | 2, 3 | 308 | ||
| DY310329.1 | - | 2, 3 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem101| Gene Symbol: Tmem101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191650.1 | NP_001178579.1 | 2, 3 | 360 | ||
| GenBank mRNA | BC086567.1 | - | 2, 3 | 308 | ||
| DY310329.1 | - | 2, 3 | 360 | |||