siRNA Details
| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198759.1 | NP_942054.1 | 13 | 2166 | ||
| XM_006247523.2 | XP_006247585.1 | 15 | 3289 | |||
| XM_006247525.2 | XP_006247587.1 | 13 | 2662 | |||
| XM_008768290.1 | XP_008766512.1 | 15 | 3585 | |||
| XM_008768291.1 | XP_008766513.1 | 13 | 1657 | |||
| GenBank mRNA | AY394725.1 | - | 13 | 2166 | ||

Arhgap27
Genomic Map