siRNA Details
| Gene Symbol: Plekhm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009677.1 | NP_001009677.1 | 7 | 1750 | ||
| XM_006247527.2 | XP_006247589.1 | 7 | 1790 | |||
| XM_008768292.1 | XP_008766514.1 | 6 | 1570 | |||
| GenBank mRNA | BC087058.1 | - | 7 | 1750 | 7124252 | |

Plekhm1
Genomic Map