siRNA Details
| Gene Symbol: Wnt9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107055.1 | NP_001100525.1 | 4 | 786 | ||
| XM_006247529.2 | XP_006247591.1 | 4 | 1596 | |||
| XM_006247530.2 | XP_006247592.1 | 5 | 1801 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt9b| Gene Symbol: Wnt9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107055.1 | NP_001100525.1 | 4 | 786 | ||
| XM_006247529.2 | XP_006247591.1 | 4 | 1596 | |||
| XM_006247530.2 | XP_006247592.1 | 5 | 1801 | |||