siRNA Details
Gene Symbol: Map3k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107058.2 | NP_001100528.1 | 13 | 1602 | ||
XM_008768358.1 | XP_008766580.1 | 13 | 1460 | |||
GenBank mRNA | BC168979.1 | - | 13 | 1602 | 7125270 |
